OKT5
88070705, Reactivity: Anti-Suppresser/cytotoxic T cells, human, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
biological source
Mouse x Mouse Hybridoma
packaging
tube of 5 μg 88070705-DNA-5UG
pkg of vial of cells 88070705-1VL
ImmunoDon
CAF1 mouse spleen
Myeloma
P3X63Ag8
Reactivity
Anti-Suppresser/cytotoxic T cells, human
technique(s)
cell culture | mammalian: suitable
isotype
IgG2
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Mouse P3X63Ag8 x Mouse CAF1 spleen
Cell Line Description
The antibody is directed against an antigen on human peripheral T cells of the suppresser/cytotoxic subset.
Immunogen
Human peripheral blood T lymphocytes
Culture Medium
IDMEM + 2mM glutamine + 20% Foetal Bovine Serum (FBS)
Subculture Routine
Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service