Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E.Derm

88032803, equine skin, Fibroblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

E.Derm, 88032803

biological source

equine skin

growth mode

Adherent

karyotype

2n = 64

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Horse dermis

Cell Line Description

Derived from a biopsy of a 4 year old mare. These cells have a finite lifespan of a total of 40 passages. Susceptible to herpes simplex, reovirus type III, vaccinia and vesicular stomatitis viruses. Not susceptible to human adenovirus type V, coxsackie A9 and B5 and polio type II. This cell line is also known as NBL-6.

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:3 i.e. seeding at 4x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C. Finite lifespan of approximately 40 passages.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service