Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

THG3-B5-13

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., 88022402, Reactivity: Anti-NP (4-hydroxy-3 nitrophenacetyl)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

Mouse + Human Transfectoma

packaging

tube of 5 μg 88022402-DNA-5UG
pkg of vial of cells 88022402-1VL

ImmunoDon

Not applicable

Myeloma

J558L

Reactivity

Anti-NP (4-hydroxy-3 nitrophenacetyl)

technique(s)

cell culture | mammalian: suitable

isotype

IgG3 G3m(g)

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse J558L + Mouse VH gene + Human C(gamma 3) gene

Cell Line Description

This human chimeric antibody is specifically directed against the hapten-4-hydroxy-3-nitrophenacetyl. V lambda of the endogenous light chain of plasmacytoma line complements VNP of transfected heavy chain to yield specific antibody. Gene marker: gpt.

Culture Medium

DMEM + 2mM glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service