Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

VERO Cell Line from African green monkey kidney

88020401, Monkey kidney, Fibroblast-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
Pricing and availability is not currently available.

Product Name

VERO Cell Line from African green monkey kidney, 88020401

biological source

Monkey kidney

description

Monkey African Green kidney

growth mode

Adherent

karyotype

2n=60

morphology

Fibroblast-like

shipped in

dry ice

storage temp.

−196°C

Application

A WHO working cell bank of 1000 ampules deposited at ECACC. Derived from an original ATCC culture. Available free of charge to companies producing vaccines.

Culture Medium

DMEM + 2 mM L-Glutamine + 10% FBS

Other Notes

Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service