Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

COLO 320HSR

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

human colon

packaging

tube of 5 μg 87101501-DNA-5UG
pkg of vial of cells 87101501-1VL

growth mode

Suspension

karyotype

Hyperdiploid, modal no. 52

morphology

Round & Refractile

products

Serotonin, epinephrine, NPP, Adrenocorticotropic hormone (ACTH) and PTH

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian colon adenocarcinoma

Cell Line Description

Subclone of COLO320DM, characterised by marker chromosomes with homogeneously staining regions (HSR) and double minutes (DM) at very low levels.

Application

Hormone studies

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C. Will form clusters in suspension.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service