Key Documents
31F4L
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.
About This Item
UNSPSC Code:
41106514
Pricing and availability is not currently available.
biological source
African green monkey kidney
packaging
tube of 5 μg 87081901-DNA-5UG
pkg of vial of cells 87081901-1VL
growth mode
Adherent
karyotype
Not specified
morphology
Epithelial
receptors
Not specified
Cell Line Origin
Monkey African Green kidney
Cell Line Description
31F4L has been transfected with Epstein Barr virus DNA but is not considered to be transformed, i.e., does not form foci on agar and does retain epithelial morphology.
Culture Medium
DMEM (D6546) + 2 mM L-Glutamine (G7513) + 5-10% FBS / FCS (F2442).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:3 to 1:6, i.e., seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37 °C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)