Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MLA144

86102901, gibbon lymph, Lymphoblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
Pricing and availability is not currently available.

Product Name

MLA144, 86102901, from gibbon lymph, Convention on the International Trade in Endangered Species of Wild Fauna and Flora (CITES)

biological source

gibbon lymph

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblast

receptors

Not specified

relevant disease(s)

cancer

Looking for similar products? Visit Product Comparison Guide

Cell Line Origin

Gibbon lymphosarcoma

Cell Line Description

A continuous lymphoid T cell line derived from a Gibbon lymphosarcoma. Cells lack the Fc Ig receptor. Media conditioned by MLA144 supports the growth of T cells from other primates and humans. MLA 144 are positive for the Gibbon ape leukemia virus (GaLV), a simian retrovirus with a moderate oncogenic risk. Handle at ACDP Category 2 containment.

Culture Medium

RPMI 1640 (R0883)+ 2 mM L-Glutamine (G7513) + 20% FBS / FCS (F2442).

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37 °C.

Other Notes

Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service