Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

OX-45

Reactivity: Anti-BCM1, rat, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., 86100910

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

Mouse x Mouse Hybridoma

packaging

tube of 5 μg 86100910-DNA-5UG
pkg of vial of cells 86100910-1VL

ImmunoDon

BALB/c mouse spleen

Myeloma

NS0/1

Reactivity

Anti-BCM1, rat

technique(s)

cell culture | mammalian: suitable

isotype

IgG1

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse NS0/1 x Mouse BALB/c spleen

Cell Line Description

The antibody recognises a 45kDa glycoprotein expressed on all haemopoietic and endothelial cells. The antigen is identical to mouse BCM1 antigen. The antibody inhibits allogeneic mixed lymphocyte reactions using lymph node cells as responders and spleen cells as stimulators.

Immunogen

Rat T blasts prepared by stimulating purified T helper cells with allogeneic irradiated rat spleen cells

Culture Medium

RPMI 1640 + 2mM glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Seed at 5x10,000 viable cells/ml. Subculture when cells reach 5x100,000-1x1,000,000 cells/ml (usually after 3-4 days); 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service