Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

RT4/31

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

human bladder

packaging

tube of 5 μg 86082102-DNA-5UG
pkg of vial of cells 86082102-1VL

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human bladder transitional cell papilloma

Cell Line Description

Derivative of RT4 which was derived from a transitional cell papilloma of a human bladder.

Culture Medium

McCoys 5a + 2 mM Glutamine + 10% Fetal Bovine Serum (FBS)

Subculture Routine

Split confluent cultures 1:3-1:6 ie seeding at 2-4 x 10,000 cells/cm2 using 0.25% trypsin or trypsin/ EDTA ; 5%CO2; 37°C

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service