Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

JH4 clone 1

86070201, guinea pig lung, Fibroblast-like

Sign Into View Organizational & Contract Pricing

Select a Size

86070201-DNA-5UG
$402.00
1 VIAL
$847.00

$402.00


Usually ships in 1 week. (Orders outside of Europe & Scandinavian countries, allow an additional week for delivery


Select a Size

Change View
86070201-DNA-5UG
$402.00
1 VIAL
$847.00

About This Item

UNSPSC Code:
41106514

$402.00


Usually ships in 1 week. (Orders outside of Europe & Scandinavian countries, allow an additional week for delivery

Product Name

JH4 clone 1, 86070201, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

biological source

guinea pig lung

packaging

tube of 5 μg 86070201-DNA-5UG
pkg of vial of cells 86070201-1VL

growth mode

Adherent

karyotype

2n = 64, diploid

morphology

Fibroblast-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Guinea pig lung

Cell Line Description

Derived from the lung of a young guinea pig (strain 13) and subsequently cloned. Capable of at least 69 population doublings without showing overt signs of senescence.

Application

Cloning studies

Culture Medium

Ham′s F12 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:4 i.e. seeding at 3-4x10,000 cells/cm2 using 0.25% trypsin; 5% CO2; 37°C. Cultures take 3-5 days to reach confluency.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service