Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AFRC MAC 75

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., 85102260, Reactivity: Anti-Nitrate Reductase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

Rat x Rat Hybridoma

packaging

tube of 5 μg 85102260-DNA-5UG
pkg of vial of cells 85102260-1VL

ImmunoDon

F344 rat spleen

Myeloma

Y3Ag1.2.3

Reactivity

Anti-Nitrate Reductase

technique(s)

cell culture | mammalian: suitable

isotype

IgG1

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat Y3Ag1.2.3 x Rat F344 spleen

Cell Line Description

The antibody specific for NADH nitrate reductase and inhibits the reduction of nitrate to nitrite using NADH as electron donor. It also inhibits the reduction of nitrate using flavin nucleotides (FMNH2 or FADH2) and methyl viologen. The antibody is directed against NADH-NR of mono and dicotyledons.

Immunogen

Nitrate reductase purified from the leaves of 6 to 8 week old spinach plants - Spinacea oleracea l

Culture Medium

DMEM + 2mM glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C. These cells require mouse macrophage feeder layers upon resuscitation

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service