Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

50 AZA SEL

85062505, Application: Fusion partner, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

Mouse x Sheep Heterohybridoma

packaging

tube of 5 μg 85062505-DNA-5UG
pkg of vial of cells 85062505-1VL

ImmunoDon

Sheep

Myeloma

GA4 (mouse x sheep heterohybridoma)

Reactivity

Non Ig secreting

technique(s)

cell culture | mammalian: suitable

isotype

Not specified

shipped in

dry ice

storage temp.

−196°C

Cell Line Description

This heterohybridoma was produced by fusion of sheep lymphocytes with the HAT sensitive GA4 sheep x mouse heterohybridoma (85662502). 50 AZA SEL is also HAT sensitive and can be used as a fusion partner.

Immunogen

Not applicable

Application

Fusion partner

Culture Medium

RPMI 1640 + 2mM glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service