RAMOS-AW, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., 85062003
biological source
human lymph
packaging
tube of 5 μg 85062003-DNA-5UG pkg of vial of cells 85062003-1VL
Derived by in vitro infection of Ramos cells (Sigma Catalogue number. 85030802) with EBV. The line is EBV positive. The cells have B lymphocyte characteristics, with surface associated mu and kappa chains.
Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)