Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

RT112/84

85061106, human bladder, Epithelial

Synonym(s):

RT112-84 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

RT112/84, 85061106

biological source

human bladder

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human bladder carcinoma epithelial

Cell Line Description

A slow growing tumour which does not form colonies in soft agar. Tumorigenic in nude mice, RT1112/84 cells possess a low plating efficiency requiring 400 cells/dish to form any colonies.

Application

RT112/84 cancer cell line has been used to study the cytotoxic effect of lipoic acid and calcium hydroxycitrate concentrations on RT112/84 cells.[1]
Tumourigenicity studies

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 10,11
D13S317: 13,14
D16S539: 11,13
D5S818: 10,13
D7S820: 11,12
THO1: 7
TPOX: 8,11
vWA: 14,17

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) seeding at 2-4x10,000 cells/cm2 using 0.05% trypsin/EDTA; 5% CO2; 37°C. Subculture weekly - cells are difficult to trypsinise, and should be kept sub-confluent.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service