AFRC MAC 54
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., 85060407, Reactivity: Anti-Phytochrome
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
biological source
Rat x Rat Hybridoma
packaging
tube of 5 μg 85060407-DNA-5UG
pkg of vial of cells 85060407-1VL
ImmunoDon
AO rat spleen
Myeloma
YB2/3.0 Ag20
Reactivity
Anti-Phytochrome
technique(s)
cell culture | mammalian: suitable
isotype
IgG2a
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Rat YB2/3.0 Ag20 x Rat AO spleen
Cell Line Description
The antibody is directed against phytochrome from Avena. It fails to distinguish the red-absorbing and the far-red-absorbing forms of phytochrome.
Immunogen
Phytochrome, purified from dark-grown Avena Sativa l seedlings
Culture Medium
DMEM + 2mM glutamine + 10% Foetal Bovine Serum (FBS)
Subculture Routine
Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C. These cells require mouse macrophage feeder layers upon resuscitation
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service