Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MCF7/AnaR-4

16022519, human breast, Polygonal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

MCF7/AnaR-4, from human breast, 16022519

biological source

human breast

growth mode

Adherent

morphology

Polygonal

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

General description

This breast cancer cell is oestrogen receptor positive and line resistant to the aromatase inhibitor anastrozole .

Cell Line Description

The MCF7/AnaR-4 cell Line was developed as a model of resistance to anti-cancer treatment with aromatase inhibitors. This breast cancer cell line resistant to the aromatase inhibitor anastrozole and is oestrogen receptor positive.
Derivation: Anastrozole-resistant cell lines were established from MCF7 cells grown in medium with 10% newborn calf serum (NCS) and 10-7 Molar testosterone. A culture of MCF7 cells were treated with 10-7 Molar anastrozole for one week, trypsinized and seeded in serial dilutions in 24-well plates. Single colonies were transferred to new wells and gradually expanded in medium with anastrozole. After ~2–3 months, the isolated colonies gave rise to anastrozole-resistant cell lines, which could be grown in anastrozole-containing medium with a weekly split ratio of ~1:25. The MCF7 cell line was authenticated in January 2014 by DNA profiling using short tandem repeat loci and found to be matching the genetic profile reported for the MCF7 cell line.

Culture Medium

Phenol red free DMEM:F12 (1:1) (D6434) + 10% Newborn Calf Serum(NBCS) (N4637) + 2.5 mM Ala-Gln (G8541) + 6 ng/ ml Insulin (I9278), 0.1 uM testosterone and 0.1 uM anastrozole

Subculture Routine

Split sub-confluent cultures (70-80%). The MCF-7/AnaR-2 cell Line is sub-cultured once a week with a split ratio of approximately 1:25 and growth medium changed every second or third day. New culture are seeded at 2-4 x 10,000 cells/cm² using 0.05% trypsin or trypsin or 0.05%trypsin/EDTA solution; 5% CO2; 37°C. Recommend media changes every 2-3 days

Other Notes

Cultures from PHE Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Citation

If use of this culture results in a scientific publication, it should be cited in the publication as: MCF7/AnaR-4 (ECACC 16022519)

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service