Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MCF7/TAMR-7

16022509, human breast, Polygonal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

MCF7/TAMR-7, from human breast, 16022509

biological source

human breast

growth mode

Adherent

morphology

Polygonal

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Description

The MCF7/TAMR-7 Cell line is a breast cancer cell line resistant to tamoxifen. Tamoxifen is by far the most widely used drug for hormone-dependent breast cancer. However, primary or acquired resistance to Tamoxifen severely limits its clinical effectiveness. The MCF7/TAMR-7 cell line is a good model cell to study the signalling pathways, which are the major drivers of tamoxifen-resistant growth.
The parental cell line for the MCF7/TAMR-7 cells is MCF7/S0.5, which was adapted to grow at low serum concentration in order to study the effect of estradiol and tamoxifen. MCF7/TAMR-7 has been established from a clone of cells that survived long term treatment with 1 μM tamoxifen. Tamoxifen-resistant cells are passaged continuously in presence of 1 μM tamoxifen, which is lethal for the parental MCF7/S0.5 cell line.

Culture Medium

Phenol red free DMEM:F12 (1:1) (D6434) + 1% FBS (F2442), 2.5 mM Ala-Gln (G8541) + 6 ng/ml insulin. To maintain high-level resistance, medium was supplemented with Tamoxifen (1 uM).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4 x 10, 000 cells/cm² using 0.05% trypsin/EDTA; 5% CO2; 37°C. Recommend media changes every 2-3 days.

Other Notes

Cultures from PHE Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Citation

If use of this culture results in a scientific publication, it should be cited in the publication as: MCF7/TAMR-7 (ECACC 16022509)

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service