Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MCF7/182R-6

16022506, human breast, Polygonal epithelia

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

MCF7/182R-6, 16022506, from human breast

biological source

human breast

growth mode

Adherent

morphology

Polygonal epithelia

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

General description

Oestrogen receptor only. Upon withdrawal of fulvestrant, the resistant phenotype is stable and the cells express ER alpha, although at a reduced level. The MCF7/182R-6 cells do not express progesterone receptor. The MCF7/182R-6 cells express increased level of EGFR, phosphorylated EGFR and phosphorylated ErbB3 and reduced level of ErbB4 compared to the parental MCF7/S0.5 cells

Cell Line Description

The MCF7/182R-6 Cell line is a breast cancer cell line resistant to fulvestrant. The MCF7/182R-6 cell line has been established from a clone of MCF7/S0.5 cells surviving long term growth with the pure steroidal antiestrogen ICI 182,780 in 100 nM concentration, see Lykkesfeldt et al (1995). The MCF7/182R-6 cells are grown in the presence of fulvestrant.

Culture Medium

Phenol red free DMEM:F12 (1:1) (D6434) + 1% FBS (F2442), 2.5 mM Ala-Gln (G8541) and 6 ng/ml insulin. Supplemented with 100nM fulvestrant to maintain resistance.

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4 x 10, 000 cells/cm² using 0.05% trypsin or trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Cultures from PHE Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Citation

If use of this culture results in a scientific publication, it should be cited in the publication as: MCF7/182R-6 (ECACC 16022506)

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service