Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CHO-K1.Cl-0235

15042908, hamster ovary, Polygonal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

CHO-K1.Cl-0235, from hamster ovary, 15042908

biological source

hamster ovary

description

CHO-K1, non-human primate FcγRIIIa, Fc gamma receptor 3A, Pan troglodytes, chimpanzee

growth mode

Adherent

morphology

Polygonal

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Stably transfected with a pcDNA3.1/Hygro(+) based vector with a cDNA insert that had been modified to enable expression of chimpanzee FcγRIIIa as a GPI linked receptor

Cell Line Description

Clone of CHO-K1 cells expressing chimpanzee (Pan troglodytes) FcγRIIIa anchored to glycosyl phosphatidylinositol. (predicted from XM_001174057.1 sequence)

Culture Medium

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6. Seeding at 2-5 x 10,000 cells/cm2 using 0.05% trypsin/EDTA; culture conditions 5% CO2/95%air; 37oC

Other Notes

Cultures from PHE Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.
If use of this culture results in a scientific publication, it should be cited in the publication as: CHO-K1.Cl-0235 (ECACC 15042908)

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service