Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ULK1 KO MEF (SIM)

14050806, mouse embryo, Fibroblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

ULK1 KO MEF (SIM), from mouse embryo, 14050806

biological source

mouse embryo

description

Mouse embryonic fibroblast, autophagy, Unc51-like kinase, spontaneously immortalised

growth mode

Adherent

morphology

Fibroblast

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Description

Mouse embryonic fibroblasts (MEFs) derived from wild-type littermate to the ULK1-knockout mouse. MEFs that were isolated and cultured were immortalised by a standard serial passaging protocol.

Culture Medium

DMEM + 10% FCS +2mM Glutamine

Subculture Routine

Trypsinize with 0.05% Trypsin/EDTA and inoculate new cultures at 2 x 104 per cm2. Cultures must be incubated in a humidified 5% CO2/95% air incubator at 37oC.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Cultures from PHE Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service