Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

OX-117

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

Mouse x Mouse Hybridoma

packaging

tube of 5 μg 13082233-DNA-5UG
pkg of vial of cells 13082233-1VL

ImmunoDon

BALB/c mouse spleen

Myeloma

NS1

Reactivity

Human signal regulatory protein gamma (SIRPgamma)

technique(s)

cell culture | mammalian: suitable

isotype

IgG1

shipped in

dry ice

Storage temp.

−196°C

Cell Line Description

The hybridoma cell line OX-117 secretes monoclonal IgG1 antibodies reactive with human signal regulatory protein gamma but it cross reacts with SIRPalpha and SIRPbeta.

Immunogen

Recombinant human signal regulatory protein gamma (SIRPgamma)

Application

Detection of SIRPgamma (but note cross reaction with SIRPalpha and SIRPbeta).

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Fetal Bovine Serum (FBS)

Subculture Routine

Seed at 5 x 104 cells/ml (5 x 105 cells/ml at resuscitation). Subculture when cells reach 5 x 105 - 1 x 106 cells/ml (usually after 3-4 days), 5% CO2, 37°C. 20% FBS can be used at resuscitation.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Cultures from PHE Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service