Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

FM-3.26

13012554, human skin, fibroblast-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

FM-3.26, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., from human skin, 13012554

biological source

human skin

description

Human melanoma, metastasis

packaging

tube of 5 μg 13012554-DNA-5UG
pkg of vial of cells 13012554-1VL

growth mode

Adherent

morphology

fibroblast-like

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Description

FM-3.26 (ESTDAB-009) was established from a metastatic tumour of a 70-year old female patient. The FM-3.26 cell line is one of more than 170 characterized melanoma cell lines collected during the ESTDAB project (European Searchable Tumour Line Database) part of the European Commissions fifth framework infrastructures programme (contract no. QLRI-CT-2001-01325). ESTDAB contains information on HLA genotype and surface expression of HLA molecules, expression of tumour antigens, antigen processing capability, production of and response to cytokines and chemokines, apoptosis regulation and expression of adhesion/accessory molecules. Such information allows researchers to identify cells possessing specific parameters important for studies of immunity, immunogenetics, gene expression, metastasis, response to chemotherapy, and other tumour biological experimentation.

DNA Profile

STR-PCR Data:

Amelogenin: X
CSF1PO: 11,13
D13S317: 12
D16S539: 12,13
D5S818: 12
D7S820: 10
THO1: 6,9.3
TPOX: 8,11
vWA: 15,16

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Fetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) seed new cultures at 2-4 x 10,000 cells/cm² using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Cultures from PHE Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service