UKRV-Mel-14b
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.
About This Item
biological source
human lymph node
packaging
tube of 5 μg 13012529-DNA-5UG
pkg of vial of cells 13012529-1VL
growth mode
Adherent
morphology
Epithelial
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
−196°C
Cell Line Description
DNA Profile
Amelogenin: X
CSF1PO: 12
D13S317: 10,12
D16S539: 11
D5S818: 11,12
D7S820: 8,10
THO1: 7,9
TPOX: 8,9
vWA: 15,17
Culture Medium
Subculture Routine
Other Notes
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service