Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C106

12022906, human colon, Epithelial

Synonym(s):

C 106, C-106

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

C106, from human colon, 12022906

biological source

human colon

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

Cell Line Origin

Human colorectal adenocarcinoma, moderately well differentiated, Dukes′ stage A

Cell Line Description

The C106 cell line was established from a 78-year old female patient with a moderately well differentiated adenocarcinoma of the rectum classified as Dukes′ stage A.

DNA Profile

STR-PCR Data:
Amelogenin: X
CSF1PO: 10,11
D13S317: 8,14
D16S539: 12,13
D5S818: 12
D7S820: 8,10
THO1: 6
TPOX: 10,11
vWA: 18

Culture Medium

Iscove′s Modified Dulbecco′s Medium, + 10% Foetal Bovine Serum (FBS) + 2 mM Glutamine

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm² using 0.05% trypsin or trypsin/EDTA; 5% CO2; 37 °C. C106 cells grow very slowly (see growth curve); following resuscitation or subculture the cells take at least 48 hours

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service