Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C80

12022904, human colon, Epithelial

Synonym(s):

C 80, C-80

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

C80, from human colon, 12022904

biological source

human colon

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human colorectal adenocarcinoma, moderately well differentiated, Dukes′ stage D

Cell Line Description

The C80 cell line was established from a 69-year old male patient with a moderately well differentiated adenocarcinoma of the rectum classified as Dukes′ stage D.

DNA Profile

STR-PCR Data:
Amelogenin: X,Y
CSF1PO: 10,12
D13S317: 8,11
D16S539: 9,10
D5S818: 11,12
D7S820: 10,11
THO1: 6,7
TPOX: 8,12
vWA: 14,18

Culture Medium

Iscove′s Modified Dulbecco′s Medium, + 10% Foetal Bovine Serum (FBS) + 2 mM Glutamine

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6, i.e., seeding at 2-4x10,000 cells/cm2 using 0.05% trypsin or trypsin/EDTA; 5% CO2; 37 °C. C80 cells grow very slowly (see growth curve); following resuscitation or subculture the cells take at least 48 hours.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service