Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

U6A Cell line human

12021507, human fibrosarcoma, epithelial

Synonym(s):

2fTGH, 2fTGH-U6A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

U6A Cell line human, 12021507

biological source

human fibrosarcoma

growth mode

adherent

morphology

epithelial

shipped in

dry ice

storage temp.

−196°C

Looking for similar products? Visit Product Comparison Guide

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service