Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

U4C cell line human

12021505, human fibrosarcoma, Epithelial

Synonym(s):

2fTGH, 2fTGH-U4C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

U4C cell line human, 12021505

biological source

human fibrosarcoma

growth mode

Adherent

morphology

Epithelial

shipped in

dry ice

storage temp.

−196°C

Looking for similar products? Visit Product Comparison Guide

Cell Line Description

The human sarcoma cell line HT 1080 (ECACC Catalogue number 85111505) was transfected with a vector encoding a selectable marker (guanine phosphoribosyltransferase) regulated by alpha interferon to create the 2fTGH cell line, enabling the selection of mutations in genes encoding components of the interferon signalling pathway. Chemical mutagenesis of the parent 2fTGH cell line enabled the isolation of a number of IFNg signalling mutants available from ECACC: U2A (12021502), U3A (12021503), U4A (12021504), U4C (12021505), U5A (12021506), U6A (12021507). Each contains a different mutation in the IFN signalling pathway. The 2fTGH panel provides a tool for the in vitro study and comparison of disrupted interferon signalling at multiple points across the IFN pathway. The U4C mutant lacks the JAK1 kinase. U4C is unresponsive to interferon alpha and interferon gamma; furthermore it is partially defective in response to double stranded RNA.

DNA Profile

STR-PCR Data:

Amelogenin: X,Y
CSF1PO: 12
D13S317: 12,14
D16S539: 9,12
D5S818: 11,13
D7S820: 9,10
THO1: 6
TPOX: 8
vWA: 14,19

Culture Medium

DMEM + 2mM Glutamine + 10% Fetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:4 to 1:8 i.e. seeding at 1-4 x 10,000 cells/cm² using 0.05% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service