Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Me11-TG6

11120838, rat lung, Epithelial and spindle shaped biphasic morphology

Synonym(s):

Me11, TG6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

Me11-TG6, 11120838, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

biological source

rat lung

packaging

tube of 5 μg 11120838-DNA-5UG
pkg of vial of cells 11120838-1VL

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial and spindle shaped biphasic morphology

receptors

Not specified

relevant disease(s)

metastasis

Looking for similar products? Visit Product Comparison Guide

Cell Line Origin

Rat transplantable mesothelioma asbestos, primary tumour

Cell Line Description

The Me-series of mesotheliomas were generated by an intrapleural inoculation of UICC crocidolite to induce primary mesotheliomas in syngenic PVG/c Norweigan Hooded rats. Solid fragments of the tumours were transplanted and maintained by s.c. serial passage. The cell lines are metastatic in a syngeneic host.

DNA Profile

Not specified

Culture Medium

DMEM (D6546)+ L-Glutamine (G7513) + 10% FBS / FCS (F2442).

Subculture Routine

Split subconfluent cultures (70-80%) 1:4 to 1:10 using 0.25% trypsin/EDTA; 5% CO2; 37 °C. Suggested seeding density 2-4 x 10,000 cells/cm2.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service