Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MM603

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Synonym(s):

MM 603

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

human skin

packaging

tube of 5 μg 11072801-DNA-5UG
pkg of vial of cells 11072801-1VL

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

metastasis

shipped in

dry ice

Looking for similar products? Visit Product Comparison Guide

Cell Line Origin

Human melanoma metastatic site

Cell Line Description

MM603 is a human melanoma derived from a metastatic site. Cell Bank Australia reference number CBA-1359

Culture Medium

RPMI 1640 (R0883) (with 2 mM L-L-Glutamine (G7513) + 25 mM HEPES (H0887) + 10% FBS / FCS (F2442).

Subculture Routine

Optimal split ratio 1:4 (seeding density 1.2 x 104 cells/cm2). Cells incubated at 37 °C with 5% CO2. Harvest the cells using 0.05% Trypsin/EDTA at 37 °C for 5 min.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments.  Please check with your local Customer Service representative for more information.
Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service