NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., Reactivity human glycophorin A (CD235a) )
Rat hybridoma producing antibody against human glycophorin A CD235a. Cambridge Enterprise Reference number AB66:www.enterprise.cam.ac.uk.
Immunogen
Human peripheral blood mononuclear cells
Application
Not specified
Culture Medium
Iscoves Modified Dulbeccos Medium (IMDM) + 2 mM glutamine + 5% Fetal Bovine Serum (FBS). Where the cells are grown under static conditions, 0.02% EDTA, trypsin or mechanical agitation maybe required to dislodge them from the plastic.
Subculture Routine
Maintain cultures between 3-9 x 105 cells/ml. 5% CO2; 37 °C. Saturation density 9 x 105/ml
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Disclaimer
This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.