Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

JIM3

10081204, human blood, Small spherical

Synonym(s):

JIM, JIM 3, JIM-3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

JIM3, 10081204, from human blood

biological source

human blood

growth mode

Suspension

karyotype

Not specified

morphology

Small spherical

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

Cell Line Origin

Human myeloma, cancer

Cell Line Description

JIM3 is a myeloma tumour line demonstrating typical plasma cell phenotype (CD38, PCA-1 8A and CD24 positive), deficient in DNA repair. JIM3 was derived from plasma myeloma cells derived from pleural fluid of advanced multiple myeloma patient, homoplastic with JIM1. Used for in vitro study and comparison of genetic instability in myeloma tumour lines (homoplastic with JIM1) and the in vitro study of effects of DNA repair deficiency in myeloma tumour lines

DNA Profile

Not specified

Culture Medium

Dexter culture medium, consisting of Fischer′s medium + 20% FCS +10-7 M hydrocortisone sodium succinate

Subculture Routine

Seed at 2-9 x 105 cells/cm3 5% CO2; 37 °C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service