Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

TO14

10081203, human ovary, epithelial; stellate, producing hemicystic colonies which detach as spheres.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

TO14, 10081203, from human ovary

biological source

human ovary

growth mode

Adherent

karyotype

For modal chromosome numbers see Langdon SP et.al. (1988) Cancer Research Nov 1; 48(21):6166-72.

morphology

Epithelial; stellate in shape and producing hemicystic colonies which grow upwards from the substrate and detach as spheres.

products

Oestrogen receptor negative cell line.

receptors

Oestrogen receptor negative cell line. For antigen expression see Langdon SP et.al. (1988) Cancer Research Nov 1; 48(21):6166-72.

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

metastasis

shipped in

dry ice

Cell Line Origin

Human ovarian cancer; oestrogen receptor negative, drug efficacy and toxicity

Cell Line Description

TO14 is an adherent cell line derived from a metastasis to the omentum obtained at hysterectomy from a patient with a well differentiated serous adenocarcinoma. TO14 exhibits poor growth in semi-solid medium (agar). TO14 is from the same patient as PEO23 and PEO14 cell lines (Sigma catalog numbers 10032313 and 10032311 respectively).

This cell line is one of nine from the PE ovarian adenocarcinoma panel (derived from 4 patients at varying stages of ovarian cancer, isolated from various malignant sites, and at various treatment stages) available at ECACC which provides a model system for research into the mechanism of oestrogen action on ovarian adenocarcinoma tumour cells, and for the study of efficacy and toxicity of oestrogen protagonists.

DNA Profile

Not specified

Culture Medium

RPMI 1640 + 2 mM Glutamine + 2 mM Sodium Pyruvate + 10% Fetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:4 to 1:10 seeding at 2- 3 x 104 cells/cm2<sup/>

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service