Mouse monoclonal against human CD-120b. TNF receptor p75 prepared from U937 cells (utr-mAb series).
Immunogen
Human CD120b (TNF receptor p75)
Application
Sandwich ELISA for soluble p75 TNF-R.
Culture Medium
Iscoves Modified Dulbeccos Medium (IMDM) + 2 mM glutamine + 25 U/ml Interleukin-6 (IL-6) + 5-10% Fetal Bovine Serum (FBS). Where the cells are grown under static conditions, 0.02% EDTA maybe required to dislodge them from the plastic.
Subculture Routine
Seed cultures between 1-3 x 105 cells/ml; Saturation density 8-10 x 105 5% CO2; 37 °C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.