Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PEO6

10032310, human ovary, Epithelial; islands of uniform polygonal cells.

Synonym(s):

PE06

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

PEO6, 10032310, from human ovary

biological source

human ovary

growth mode

Adherent

karyotype

For modal chromosome numbers see Langdon SP et.al. (1988) Cancer Research Nov 1; 48(21):6166-72.

morphology

Epithelial; islands of uniform polygonal cells.

products

Not specified

receptors

Oestrogen receptor positive cell line. For antigen expression see Langdon SP et.al. (1988) Cancer Research Nov 1; 48(21):6166-72.

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

Cell Line Origin

Human ovarian cancer; oestrogen receptor positive,

DNA Profile

STR-PCR Data:
Amelogenin: X
CSF1PO: 10,12
D13S317: 10
D16S539: 9
D5S818: 11,12
D7S820: 10
THO1: 9.3
TPOX: 9,11
vWA: 15,16

Culture Medium

RPMI 1640 + 2 mM Glutamine + 2 mM Sodium Pyruvate + 10% Fetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:4 to 1:10 seeding at 2- 3 x 104 cells/cm2 using 0.05% trypsin or trypsin/EDTA; 5% CO2; 37 °C. Doubling time approximately 77 hours.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service