CMT 167 (clone of CMT 64) from mouse lung, 10032302
biological source
Mouse strain C57BL/lcrf lung
description
Lung carcinoma from mouse
growth mode
Adherent
karyotype
Not specified
morphology
Epithelial, closely packed sheets at confluence
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
metastasis
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Lung carcinoma from mouse
Mouse lung carcinoma; metastasis
Cell Line Description
CMT 167 is a highly metastatic subclone of murine alveogenic lung carcinoma cell line CMT 64 (Sigma Catalogue number 10032301). CMT 167 was isolated by subcloning and in vivo screening for high metastatic potential.
Application
For complete product information, please see ECACC
Split sub-confluent cultures (70-80%) 1:4 to 1:10 seeding at approximately 3 x 104 cells/cm2 using 0.05% trypsin or trypsin/EDTA; 5% CO2; 37°C. Saturation density 1.2 x 105 cells/cm2
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Disclaimer
This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.