Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HRA-19a1.1 Alpha2Alpha1 B

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

human colon

packaging

tube of 5 μg 09071524-DNA-5UG
pkg of vial of cells 09071524-1VL

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human colorectal adenocarcinoma

Cell Line Description

Genetically modified HRA-19a1.1 cells (Sigma catalog number 09071519) expressing a chimeric alpha2 integrin with the extracellular and transmembrane domain of alpha2 integrin and the cytoplasmic domain of alpha1 integrin.

DNA Profile

STR-PCR Data:

Amelogenin: X,Y
CSF1PO: 12
D13S317: 11,12
D16S539: 11
D5S818: 11
D7S820: 9,12
THO1: 8
TPOX: 11
vWA: 16

Culture Medium

DMEM (High glucose Sigma D6546) + 2 mM Glutamine + 10% Fetal Bovine Serum (FBS) + G418 200 μg/ml.

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:4 weekly using 0.05% trypsin/EDTA; 7.5% CO2; 37 °C. Suggested seeding density 2-4x10,000 cells/cm2.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service