Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

UM-RC-2

8090511, human kidney, Polygonal

Synonym(s):

Human UM-RC-2, UM-RC-2 Cell Line, Vector UM-RC-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

UM-RC-2, 08090511

biological source

human kidney

growth mode

Adherent

karyotype

Median number of chromosomes is 57

morphology

Polygonal

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human renal carcinoma

Cell Line Description

UM-RC-2 is a continuous cell line established from a primary renal adenocarcinoma. It exhibits a loss of contact inhibition and produces tumours in the nude mouse model. The patient had metastatic disease at the time of their nephrectomy.

Application

Study of tumour associated antigens

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 1-2x10,000 cells/cm2 using 0.05% trypsin/EDTA; 5% CO2; 37°C. Doubling time approximately 24 hours.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service