Recommended Products
Product Name
UM-UC-4, 08090501
biological source
human lymph
growth mode
Adherent
karyotype
Modal number of chromosomes is 51
morphology
Epithelial
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
metastasis
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human adenocarcinoma lymph node metastasis
Cell Line Description
UM-UC-4 was obtained from an open operative specimen from a metastatic tumour of the lymph node; the female patient had an urachal carcinoma. UM-UC-4 was found to produce tumours in athymic nude mice.
DNA Profile
STR-PCR Data:
Amelogenin: X
CSF1PO: 11
D13S317: 9
D16S539: 13
D5S818: 11
D7S820: 9,13
THO1: 9.3
TPOX: 8
vWA: 17,19
Amelogenin: X
CSF1PO: 11
D13S317: 9
D16S539: 13
D5S818: 11
D7S820: 9,13
THO1: 9.3
TPOX: 8
vWA: 17,19
Culture Medium
EMEM (EBSS) + 2 mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Fetal Bovine Serum (FBS).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:4 to 1:8, i.e., seeding at 2-4x10,000 cells/cm2 using 0.05% trypsin/EDTA; 5% CO2; 37 °C. Doubling time approximately 32 hours.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Disclaimer
This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service