Pricing and availability is not currently available.
Product Name
108CC5T-BU-1, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., 08062570
biological source
Mouse x Rat Hybridoma nerve
packaging
tube of 5 μg 08062570-DNA-5UG pkg of vial of cells 08062570-1VL
A bromodeoxyuridine resistant mutant of 108CC5 (Sigma Catalogue number 08062521), a mouse neuroblastoma N18TG2 x rat glioma C6-BU-1 hybrid.
Application
Used in the generation of cell hybrids.
Culture Medium
DMEM + 2 mM glutamine + 10% Foetal Bovine Serum (FBS) + 1 mM 8-bromodeoxyuridine. The cells can be grown in the absence of bromodeoxyuridine.
Subculture Routine
Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 1-2x10,000 cells/cm2 using 0.05% trypsin/EDTA; 5% CO2; 37°C. The cultures rapidly produce lactic acid through anaerobic glycolysis causing the media to become acidified. The medium must never be allowed to become acidic as cells detach in clumps and take many passages to recover. Frequent inspection of cultures is therefore required. Immediately after resuscitation a pre-centrifugation step to remove cryoprotectant (DMSO) is recommended i.e. pellet the cells by centrifugation at 100 x g for 5 minutes and resuspend the cell pellet in fresh medium using the appropriate volume to achieve the correct seeding density.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.