Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B82 (Hamprecht)

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

mouse connective tissue

packaging

tube of 5 μg 08062522-DNA-5UG
pkg of vial of cells 08062522-1VL

growth mode

Adherent

karyotype

Modal No of chromosomes is 53

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse Fibroblast (L cells)

Cell Line Description

An 8-bromodeoxyuridine resistant mutant of mouse L cell fibroblasts (Mouse strain C3H/An). B82 lacks thymidine kinase and was isolated by virtue of resistance to 8-bromodeoxyuridine (30 μg/ml). ECACC has two catalogue entries for the B82 cell line which differ in that they were deposited by different groups: B82 (Sigma Catalogue number 85011408) and B82(Hamprecht) (Sigma Catalogue number 08062522). Several other cell lines in the ECACC catalogue have been derived from B82(Hamprecht).

Application

Used in the generation of cell hybrids.

Culture Medium

DMEM + 2 mM glutamine + 10% Foetal Bovine Serum (FBS) + 1 mM 8-bromodeoxyuridine

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.05% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service