Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NS20Y-BU-7

8062510, mouse nerve, Neuronal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

NS20Y-BU-7, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA., 08062510

biological source

mouse nerve

packaging

tube of 5 μg 08062510-DNA-5UG
pkg of vial of cells 08062510-1VL

growth mode

Adherent

karyotype

Not specified

morphology

Neuronal

products

Choline acetyltransferase

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse neuroblastoma

Cell Line Description

A bromodeoxyuridine-resistant mutant of NS20Y (Sigma Catalogue number 08062517) The cell line NS20Y is a cholinergic cell line derived from mouse neuroblastoma C1300 Mouse strain A/Jax.

Application

Used in the generation of cell hybrids.

Culture Medium

DMEM + 2 mM Glutamine + 10% Foetal Bovine Serum (FBS)+ 1 mM 8-bromodeoxyuridine. Cells can also be grown in the absence of bromodeoxyuridine

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.05% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service