TGFbeta type I receptor, TGFbeta II receptor, TGFbeta III receptor
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human oral squamous cell carcinoma, buccal mucosa
Cell Line Description
Established from a squamous cell carcinoma (SCC) of the buccal mucosa (20mm-40mm) of a 53 year-old female patient. STNMP stage II, moderately differentiated, node negative tumour. Mutant p53, codon 68 exon 4, G to T; wild type K-, N- and Ha-ras. These cells are non-tumourigenic on subcutaneous injection into athymic nude mice, but tumourigenic on injection into the floor of the mouth.
Split sub-confluent cultures (70-80%), approximately every 5-6 days, 1:8 to 1:10 using 0.05% trypsin/EDTA; 5% CO2; 37°C. Suggested seeding density 5 x 1000 cells/cm2. Cells can take approximately 10 minutes to detach, an alternative is to trypsinise 2 to 3 times with fresh trypsin for shorter periods for each trypsin application. Avoid knocking flasks during the trypsinisation process as this can lead to loss of viability.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Disclaimer
This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.