TGFbeta type I receptor, TGFbeta II receptor, TGFbeta III receptor
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human oral squamous cell carcinoma, floor of mouth
Cell Line Description
Established from a squamous cell carcinoma (SCC) of the floor of the mouth (20 mm-40 mm) of a 40 year-old female patient. STNMP stage III, well differentiated, node positive tumour. Mutant p53, codon 266 exon 8, G to T; wild type K-, N-, and Ha-ras. This cell line is highly responsive to TGF (Transforming growth factor )-β. H376 cells in culture scatter, i.e., the cells redistribute on the substrate, in response to Hepatocyte Growth Factor. Non-tumourigenic in athymic nude mice by subcutaneous injection and on injection into the floor of the mouth (orthotopic). Haplotype information: A*03,A*24; B*27,B*35; Cw*04,Cw*12
Split sub-confluent cultures (70-80%), approximately every 5-6 days, 1:8 to 1:10 using 0.05% trypsin/EDTA; 5% CO2; 37 °C. Suggested seeding density 5 x 1000 cells/cm2.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Disclaimer
This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.