Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H314

6092003, human mouth (floor), Polygonal & spindle

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

H314, 06092003

biological source

human mouth (floor)

growth mode

Adherent

karyotype

Not specified

morphology

Polygonal & spindle

products

Not specified

receptors

TGFbeta type I receptor, TGFbeta II receptor, TGFbeta III receptor

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human oral squamous cell carcinoma, floor of mouth

Cell Line Description

Established from a squamous cell carcinoma (SCC) of the floor of the mouth (20mm-40mm) of an 82 year-old male patient. STNMP stage II, moderately differentiated, node positive tumour. Mutant p53, codon 176 exon 5, G to T and 373 exon 11, A to G; wild type K-, N- and Ha-ras. Tumourigenic in athymic nude mice on subcutaneous injection; on injection into the floor of the mouth, cells spread to lymphatics and lymph nodes. By short tandem repeat (STR)-PCR analysis the Y chromosome could not be detected in this cell line when tested at ECACC. It is a known phenomenon that SSC cell lines can lose their Y chromosome.

Culture Medium

DMEM:HAMS F12 (1:1) + 2mM Glutamine + 10% Foetal Bovine Serum (FBS) + 0.5 ug/ml sodium hydrocortisone succinate

Subculture Routine

Split sub-confluent cultures (70-80%), approximately every 6-7 days, 1:8 to 1:10 using 0.05% trypsin/EDTA; 5% CO2; 37°C. Suggested seeding density 7 x 1000 cells/cm2.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service