Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H103

6092001, human mouth, Polygonal

Synonym(s):

H 103

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

H103, 06092001

biological source

human mouth

growth mode

Adherent

karyotype

Aneuploid

morphology

Polygonal

products

Not specified

receptors

TGFbeta type I receptor, TGFbeta II receptor, TGFbeta III receptor

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human oral squamous cell carcinoma, tongue

Cell Line Description

Established from a small squamous cell carcinoma (SSC) of the tongue (<20mm in size) of a 32 year-old male patient. STNMP stage 1, well differentiated, node negative tumour. Aneuploid- dual G-/G1 peak on cell cycle analysis by flow cytometry. Mutant p53, codon 244 exon 7; G to T; wild type K-, N- and Ha-ras. Tumourigenic in athymic nude mice, both by subcutaneous injection and injection into the floor of the mouth. By short tandem repeat (STR)-PCR analysis the Y chromosome could not be detected in this cell line when tested at ECACC. It is a known phenomenon that SSC cell lines can lose their Y chromosome.

Culture Medium

DMEM:HAMS F12 (1:1) + 2mM Glutamine + 10% Foetal Bovine Serum (FBS) + 0.5 ug/ml sodium hydrocortisone succinate

Subculture Routine

Split sub-confluent cultures (70-80%) 1:8 to 1:10 using 0.05% trypsin/EDTA; 5% CO2; 37°C. Suggested seeding density 5 x 1000 cells/cm2. Cells can take approximately 10 minutes to detach, an alternative is to trypsinise 2 to 3 times with fresh trypsin for shorter periods for each trypsin application. Avoid knocking flasks during the trypsinisation process as this can lead to loss of viability.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service