1156QE/8
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
biological source
human embryo (carcinoma)
packaging
tube of 5 μg 06072609-DNA-5UG
pkg of vial of cells 06072609-1VL
growth mode
Not specified
karyotype
Not specified
morphology
Not specified
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
Cell Line Origin
Human embryonal carcinoma
Cell Line Description
A human embryonal carcinoma cell line established from a primary testicular tumour of a 22 year-old patient. By short tandem repeat (STR)-PCR analysis the Y chromosome could not be detected in this cell line when tested at ECACC. It is a known phenomenon that due to the increased genetic instability of cancer cell lines the Y chromosome can be rearranged or lost resulting in lack of detection.
DNA Profile
STR-PCR Data:
Amelogenin: X
CSF1PO: 11
D13S317: 8,9
D16S539: 11,14
D5S818: 11,12
D7S820: 9
THO1: 9.3
TPOX: 8
vWA: 16,17
Amelogenin: X
CSF1PO: 11
D13S317: 8,9
D16S539: 11,14
D5S818: 11,12
D7S820: 9
THO1: 9.3
TPOX: 8
vWA: 16,17
Culture Medium
DMEM (High Glucose) + 2 mM Glutamine + 10% Fetal Bovine Serum (FBS). No sodium pyruvate.
Subculture Routine
Split sub-confluent cultures (70-80%) 1:3 to 1:5 i.e. seeding at >6x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 10% CO2; 37°C. Cells need to be maintained at a high cell density.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service