Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

RaBE (Slow Growth)

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

rat lung

packaging

tube of 5 μg 04100602-DNA-5UG
pkg of vial of cells 04100602-1VL

growth mode

Adherent

karyotype

Hypotetraploid, n=74 chromosomes (median)

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat Bronchial Epithelial Cells (RaBE) Slow Growth

Cell Line Description

Spontaneously immortalised rat bronchial epithelial cell line derived from a 4 month old male Wistar rat. The cell line is basal cell-like expressing cytokeratins 14 and 15. Vimentin negative. This is a slower growing derivative of the cell line RaBE, Sigma Catalogue number. 04100601, adapted to culture medium without growth factors.

Culture Medium

Ham?s F12 + 2mM Glutamine + 5% Foetal Bovine Serum (FBS)

Subculture Routine

Split sub-confluent cultures (70-80%) between 1:100 and 1:800 i.e. seeding at 20,000 cells/25cm2 with 5ml medium or 5x100,000 cells/75cm2 with 20ml, using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C. Subculture once a week, double volume at medium change

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service