RaBE
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
biological source
rat lung
packaging
tube of 5 μg 04100601-DNA-5UG
pkg of vial of cells 04100601-1VL
growth mode
Adherent
karyotype
Hypotetraploid, n=74 chromosomes (median)
morphology
Epithelial
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Rat Bronchial Epithelial Cells (RaBE)
Cell Line Description
Spontaneously immortalised rat bronchial epithelial cell line derived from a 4 month old male Wistar rat. The cell line is basal cell-like expressing cytokeratins 14 and 15. Vimentin negative. A slow growing derivative of RaBE, cultured in the absence of growth factors is also available - RaBE (Slow Growth), Sigma Catalogue number. 04100602.
Culture Medium
Ham′s F12 + 2mM Glutamine + 10ug/ml transferrin + 5ug/ml insulin + 10ng/ml EGF + 10ng/ml aFGF + 100ng/ml cholera toxin + 5% Foetal Bovine Serum (FBS).
Subculture Routine
Split sub-confluent cultures (70-80%) between 1:100 and 1:800 i.e. seeding 5000 cells/25cm2 with 5ml or 10,000 cells/75cm2 with 20ml using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C. Subculture once a week, double volume
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service