Key Documents
COS-1-InVitrus
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.
About This Item
UNSPSC Code:
41106514
Pricing and availability is not currently available.
biological source
African green monkey kidney
packaging
tube of 5 μg 04092902-DNA-5UG
pkg of vial of cells 04092902-1VL
growth mode
Adherent
karyotype
Not specified
morphology
Fibroblast
receptors
Not specified
Cell Line Origin
Monkey African green kidney, SV40 transformed, serum-free
Cell Line Description
COS-1 cell line (Sigma catalog no. 88031701) adapted to grow in the chemically defined protein- and peptide-free InVitrus medium (Cell Culture Technologies). This cell line is substrate dependent, i.e., adherent. COS-1 was derived from CV-1 simian cells transformed by origin-defective mutant of SV40 which codes for wildtype T Ag. Possible host for propagation of pure populations of recombinant SV40 virus. The cells should be handled under laboratory containment level 2.
Culture Medium
InVitrus medium (Cell Culture Technologies http://www.cellculture.com/) + 4 mM L-Glutamine (G7513) + 5 ug/ml Insulin. When freezing cells down use 50:50 fresh culture medium:conditioned medium plus 10% DMSO (D2650).
Subculture Routine
Split semi-confluent cultures 1:3, i.e., seeding at 4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37 °C. Please note: As the cell culture medium does not contain protein it will not inactivate trypsin. Once cells have detached from the flask it is necessary to dilute the trypsin with the cell culture medium, centrifuge the cells and resuspend in fresh medium. In order to prevent syncitia formation, do not allow cells to become confluent.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)