Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BICR 82

4072112, human mouth (maxilla), Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

BICR 82, 04072112

biological source

human mouth (maxilla)

growth mode

Adherent

karyotype

Aneuploid

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Upper Jaw (Maxilla) Carcinoma

Cell Line Description

A keratinocyte cell line established from a squamous carcinoma of the upper jaw (maxilla) of a Caucasian male. Keratin and involucrin markers present. Known mutations: p53, p16 and p14ARF. Tumourigenicity in athymic mice not tested. Edington et al., 1995 must be cited and the cell line referred to by its original name.

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS) + 0.4 micrograms/ml Hydrocortisone.

Subculture Routine

Split sub-confluent cultures (70-80%) 1:4 - 1:5 using 0.05% trypsin/EDTA; 8% CO2; 37°C. Suggested seeding density 2 x 10,000 cells/cm2. Culture cells on a feeder layer of lethally-irradiated or mitomycin C-treated 3T3 feeder cells.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service